Assay Details
Gene Symbol: CKMT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022413031.1 | XP_022268739.1 | 5 - 6 | 945 | 94 | |
XM_535451.6 | XP_535451.1 | 4 - 5 | 894 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
