Assay Details
Gene Symbol: PPP1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113168.2 | NP_001106639.2 | 2 - 3 | 383 | 119 | |
GenBank mRNA | BM539680.1 | - | 2 - 3 | 270 | 119 | |
CX006826.1 | - | 2 - 3 | 493 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
