Assay Details
Gene Symbol: CIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022414726.1 | XP_022270434.1 | 2 - 3 | 273 | 162 | |
XM_535968.6 | XP_535968.2 | 2 - 3 | 271 | 162 |

More Information
Set Membership: |
![]() ![]() ![]() |
