Assay Details
Gene Symbol: NDUFB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_535753.6 | XP_535753.2 | 2 - 3 | 376 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NDUFB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_535753.6 | XP_535753.2 | 2 - 3 | 376 | 89 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |