Assay Details
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014116825.2 | XP_013972300.2 | 19 - 20 | 3216 | 67 | |
XM_014116826.2 | XP_013972301.2 | 19 - 20 | 3217 | 67 | ||
XM_014116828.2 | XP_013972303.1 | 19 - 20 | 2176 | 67 | ||
XM_014116829.2 | XP_013972304.1 | 18 - 19 | 2325 | 67 | ||
XM_014116830.2 | XP_013972305.1 | 19 - 20 | 2554 | 67 | ||
XM_014116831.2 | XP_013972306.1 | 4 - 5 | 478 | 67 | ||
XM_022423432.1 | XP_022279140.1 | 14 - 15 | 1546 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
