Assay Details
Gene Symbol: ADA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005635099.2 | XP_005635156.1 | 4 - 5 | 476 | 81 | |
XM_005635103.3 | XP_005635160.1 | 4 - 5 | 564 | 81 | ||
XM_014106890.2 | XP_013962365.1 | 4 - 5 | 463 | 81 | ||
XM_022409460.1 | XP_022265168.1 | 4 - 5 | 408 | 81 | ||
XM_022409461.1 | XP_022265169.1 | 4 - 5 | 408 | 81 | ||
XM_022409462.1 | XP_022265170.1 | 4 - 5 | 408 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
