Assay Details
Gene Symbol: RPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014111641.2 | XP_013967116.1 | 1 - 2 | 166 | 85 | |
XM_014111642.2 | XP_013967117.1 | 1 - 2 | 168 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014111641.2, XM_014111642.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
