Assay Details
Gene Symbol: DSE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022419122.1 | XP_022274830.1 | 4 - 5 | 826 | 68 | |
XM_022419123.1 | XP_022274831.1 | 4 - 5 | 719 | 68 | ||
XM_533477.6 | XP_533477.2 | 5 - 6 | 1286 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
