Assay Details
Gene Symbol: TREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022426027.1 | XP_022281735.1 | 7 - 8 | 1459 | 113 | |
XM_022426028.1 | XP_022281736.1 | 7 - 8 | 1393 | 113 | ||
XM_532133.6 | XP_532133.2 | 4 - 5 | 708 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
