Assay Details
Gene Symbol: BLVRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022425584.1 | XP_022281292.1 | 3 - 4 | 481 | 80 | |
XM_533666.6 | XP_533666.2 | 4 - 5 | 566 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
