Assay Details
Gene Symbol: ABITRAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014117909.2 | XP_013973384.1 | 3 - 4 | 966 | 142 | |
XM_532026.6 | XP_532026.2 | 4 - 5 | 408 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
