Assay Details
Gene Symbol: HECA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417566.1 | XP_022273274.1 | 1127 | 95 | ||
XM_022417569.1 | XP_022273277.1 | 1257 | 95 | |||
XM_022417573.1 | XP_022273281.1 | 1257 | 95 | |||
XM_022417577.1 | XP_022273285.1 | 1152 | 95 | |||
XM_022417578.1 | XP_022273286.1 | 1146 | 95 | |||
XM_533428.6 | XP_533428.2 | 1163 | 95 |
