Assay Details
Gene Symbol: HECA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417566.1 | XP_022273274.1 | 1285 | 77 | ||
XM_022417577.1 | XP_022273285.1 | 1310 | 77 | |||
XM_022417578.1 | XP_022273286.1 | 1304 | 77 | |||
XM_533428.6 | XP_533428.2 | 1321 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
