Assay Details
Gene Symbol: TXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014117372.2 | XP_013972847.1 | 2 - 3 | 411 | 64 | |
XM_531748.6 | XP_531748.3 | 2 - 3 | 411 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
