Assay Details
Gene Symbol: SMIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618615.3 | XP_005618672.1 | 2 - 3 | 347 | 92 | |
XM_022417229.1 | XP_022272937.1 | 2 - 3 | 349 | 92 | ||
XM_022417230.1 | XP_022272938.1 | 2 - 3 | 244 | 92 | ||
XM_022417231.1 | XP_022272939.1 | 2 - 3 | 209 | 92 | ||
XM_022417232.1 | XP_022272940.1 | 2 - 3 | 352 | 92 | ||
XM_536252.6 | XP_536252.1 | 2 - 3 | 324 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
