Assay Details
Gene Symbol: OFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005641122.3 | XP_005641179.1 | 6 - 7 | 836 | 71 | |
XM_014111611.2 | XP_013967086.1 | 5 - 6 | 463 | 71 | ||
XM_022416439.1 | XP_022272147.1 | 6 - 7 | 836 | 71 | ||
XM_537958.6 | XP_537958.3 | 6 - 7 | 679 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
