Assay Details
Gene Symbol: LAMTOR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621793.2 | XP_005621850.1 | 2 - 3 | 475 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LAMTOR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621793.2 | XP_005621850.1 | 2 - 3 | 475 | 98 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |