Assay Details
Gene Symbol: PMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014114296.2 | XP_013969771.1 | 2385 | 71 | ||
XM_022419723.1 | XP_022275431.1 | 1782 | 71 | |||
XM_536879.6 | XP_536879.3 | 2314 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
