Assay Details
Gene Symbol: RELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014112639.2 | XP_013968114.1 | 2 - 3 | 288 | 72 | |
XM_022417251.1 | XP_022272959.1 | 2 - 3 | 425 | 72 | ||
XM_536264.5 | XP_536264.2 | 2 - 3 | 423 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
