Assay Details
Gene Symbol: CMTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620875.2 | XP_005620932.1 | 3 - 4 | 673 | 137 | |
XM_536824.5 | XP_536824.3 | 3 - 4 | 668 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
