Assay Details
Gene Symbol: BCL7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022419848.1 | XP_022275556.1 | 595 | 108 | ||
XM_022419849.1 | XP_022275557.1 | 594 | 108 | |||
XM_022419850.1 | XP_022275558.1 | 591 | 108 | |||
XM_536901.6 | XP_536901.1 | 1 | 583 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
