Assay Details
Gene Symbol: SURF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625145.3 | XP_005625202.1 | 3 - 4 | 332 | 117 | |
XM_022423632.1 | XP_022279340.1 | 3 - 4 | 262 | 117 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005625145.3, XM_022423632.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
