Assay Details
Gene Symbol: CDKN2AIPNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_538628.5 | XP_538628.2 | 1 - 2 | 287 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
