Assay Details
Gene Symbol: NBAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630144.3 | XP_005630201.1 | 4346 | 78 | ||
XM_022404466.1 | XP_022260174.1 | 4398 | 78 | |||
XM_022404467.1 | XP_022260175.1 | 4398 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
