Assay Details
Gene Symbol: CANT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624035.3 | XP_005624092.1 | 1398 | 85 | ||
XM_005624037.2 | XP_005624094.1 | 595 | 85 | |||
XM_022422660.1 | XP_022278368.1 | 1560 | 85 | |||
XM_540464.5 | XP_540464.2 | 570 | 85 |
