Assay Details
Gene Symbol: NEDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629253.3 | XP_005629310.1 | 4 - 5 | 507 | 134 | |
XM_005629254.2 | XP_005629311.1 | 2 - 3 | 202 | 134 | ||
XM_005629255.3 | XP_005629312.1 | 4 - 5 | 371 | 134 | ||
XM_005629256.2 | XP_005629313.1 | 4 - 5 | 507 | 134 | ||
XM_022403357.1 | XP_022259065.1 | 2 - 3 | 333 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
