Assay Details
Gene Symbol: TRMT61B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630295.2 | XP_005630352.1 | 1 - 2 | 738 | 78 | |
XM_005630296.2 | XP_005630353.1 | 1 - 2 | 738 | 78 | ||
XM_014120492.2 | XP_013975967.1 | 1 - 2 | 738 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
