Assay Details
Gene Symbol: POLE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022404774.1 | XP_022260482.1 | 1 - 2 | 277 | 76 | |
XM_022404775.1 | XP_022260483.1 | 1 - 2 | 277 | 76 | ||
XM_540212.5 | XP_540212.3 | 1 - 2 | 277 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
