Assay Details
Gene Symbol: GABRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639456.4 | XP_003639504.2 | 2 - 3 | 369 | 81 | |
XM_022426953.1 | XP_022282661.1 | 3 - 4 | 357 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
