Assay Details
Gene Symbol: AKAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005615652.3 | XP_005615709.1 | 4 - 5 | 871 | 81 | |
XM_005615653.3 | XP_005615710.1 | 4 - 5 | 688 | 81 | ||
XM_005615654.3 | XP_005615711.1 | 4 - 5 | 863 | 81 | ||
XM_005615655.3 | XP_005615712.1 | 4 - 5 | 874 | 81 | ||
XM_005615656.3 | XP_005615713.1 | 3 - 4 | 447 | 81 | ||
XM_022420070.1 | XP_022275778.1 | 4 - 5 | 863 | 81 | ||
XM_022420076.1 | XP_022275784.1 | 4 - 5 | 861 | 81 | ||
XM_022420079.1 | XP_022275787.1 | 3 - 4 | 391 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
