Assay Details
Gene Symbol: EPM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417819.1 | XP_022273527.1 | 1196 | 94 | ||
XM_022417820.1 | XP_022273528.1 | 862 | 94 | |||
XM_022417821.1 | XP_022273529.1 | 1193 | 94 | |||
XM_022417822.1 | XP_022273530.1 | 1192 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
