Assay Details
Gene Symbol: COX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628280.3 | XP_005628337.2 | 1 - 2 | 356 | 80 | |
XM_539312.5 | XP_539312.4 | 1 - 2 | 357 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
