Assay Details
Gene Symbol: ZNF483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005626920.3 | XP_005626977.1 | 4 - 5 | 898 | 91 | |
XM_014117917.2 | XP_013973392.1 | 4 - 5 | 1099 | 91 | ||
XM_022424934.1 | XP_022280642.1 | 4 - 5 | 854 | 91 | ||
XM_022424936.1 | XP_022280644.1 | 3 - 4 | 1219 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
