Assay Details
Gene Symbol: PPIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005626445.3 | XP_005626502.1 | 4 - 5 | 555 | 80 | |
XM_022425210.1 | XP_022280918.1 | 4 - 5 | 654 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
