Assay Details
Gene Symbol: APOA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619731.3 | XP_005619788.1 | 650 | 55 | ||
XM_022418257.1 | XP_022273965.1 | 471 | 55 | |||
XM_546511.6 | XP_546511.1 | 643 | 55 |

More Information
Set Membership: |
![]() ![]() |
