Assay Details
Gene Symbol: IRAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637049.3 | XP_005637106.1 | 14 - 15 | 1222 | 113 | |
XM_005637050.3 | XP_005637107.1 | 14 - 15 | 1219 | 113 | ||
XM_005637051.3 | XP_005637108.1 | 13 - 14 | 1114 | 113 | ||
XM_543758.6 | XP_543758.2 | 13 - 14 | 1352 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
