Assay Details
Gene Symbol: ARPP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638422.3 | XP_005638479.1 | 2 - 3 | 280 | 122 | |
XM_022412585.1 | XP_022268293.1 | 3 - 4 | 274 | 122 | ||
XM_022412586.1 | XP_022268294.1 | 3 - 4 | 263 | 122 | ||
XM_022412587.1 | XP_022268295.1 | 2 - 3 | 141 | 122 | ||
XM_022412588.1 | XP_022268296.1 | 2 - 3 | 246 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
