Assay Details
Gene Symbol: EEF1AKMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005635555.3 | XP_005635612.1 | 546 | 70 | ||
XM_005635556.3 | XP_005635613.1 | 653 | 70 | |||
XM_005635557.2 | XP_005635614.1 | 644 | 70 | |||
XM_543173.5 | XP_543173.1 | 646 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
