Assay Details
Gene Symbol: PLA2G4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638288.3 | XP_005638345.1 | 4 - 5 | 708 | 81 | |
XM_014109622.2 | XP_013965097.1 | 4 - 5 | 805 | 81 | ||
XM_022412673.1 | XP_022268381.1 | 5 - 6 | 820 | 81 | ||
XM_022412674.1 | XP_022268382.1 | 4 - 5 | 709 | 81 | ||
XM_544638.6 | XP_544638.3 | 4 - 5 | 587 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
