Assay Details
Gene Symbol: AIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639250.3 | XP_005639307.1 | 3 - 4 | 394 | 145 | |
XM_005639251.3 | XP_005639308.1 | 3 - 4 | 403 | 145 | ||
XM_545016.5 | XP_545016.2 | 3 - 4 | 406 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
