Assay Details
Gene Symbol: CYB5D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_546555.5 | XP_546555.2 | 2 - 3 | 794 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CYB5D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_546555.5 | XP_546555.2 | 2 - 3 | 794 | 93 |
Set Membership: |
![]() ![]() ![]() ![]() |