Assay Details
Gene Symbol: FKBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640360.3 | XP_005640417.1 | 1 - 2 | 351 | 75 | |
XM_545546.6 | XP_545546.1 | 1 - 2 | 321 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
