Assay Details
Gene Symbol: TUBA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022410871.1 | XP_022266579.1 | 3 - 4 | 483 | 72 | |
XM_543889.5 | XP_543889.2 | 2 - 3 | 359 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022410871.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
