Assay Details
Gene Symbol: BAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637782.2 | XP_005637839.1 | 2 - 3 | 914 | 106 | |
XM_544046.5 | XP_544046.2 | 2 - 3 | 914 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
