Assay Details
Gene Symbol: TSTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022415296.1 | XP_022271004.1 | 2 - 3 | 283 | 69 | |
XM_545762.6 | XP_545762.2 | 3 - 4 | 414 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
