Assay Details
Gene Symbol: PLA2G2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022413828.1 | XP_022269536.1 | 4 - 5 | 492 | 88 | |
XM_022413830.1 | XP_022269538.1 | 4 - 5 | 625 | 88 | ||
XM_022413831.1 | XP_022269539.1 | 4 - 5 | 428 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
