Assay Details
Gene Symbol: SMUG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636748.3 | XP_005636805.1 | 3 - 4 | 669 | 64 | |
XM_022411245.1 | XP_022266953.1 | 3 - 4 | 737 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
