Assay Details
Gene Symbol: LOC489647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022419043.1 | XP_022274751.1 | 1581 | 97 | ||
XM_546767.6 | XP_546767.3 | 1683 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
