Assay Details
Gene Symbol: FUOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637912.3 | XP_005637969.1 | 2 - 3 | 207 | 119 | |
XM_005637914.3 | XP_005637971.1 | 2 - 3 | 207 | 119 | ||
XM_014109157.2 | XP_013964632.1 | 2 - 3 | 207 | 119 | ||
XM_548817.5 | XP_548817.2 | 2 - 3 | 207 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
