Assay Details
Gene Symbol: RAB7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640804.3 | XP_005640861.1 | 2 - 3 | 212 | 59 | |
XM_014111271.2 | XP_013966746.2 | 2 - 3 | 284 | 59 | ||
XM_014111272.2 | XP_013966747.2 | 3 - 4 | 438 | 59 | ||
XM_014111273.2 | XP_013966748.2 | 2 - 3 | 159 | 59 | ||
XM_014111274.2 | XP_013966749.1 | 2 - 3 | 214 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
